Assay Details
Target Gene Details
Entrez Gene ID: | 29761 |
Gene Name: | ubiquitin specific peptidase 25 |
Gene Aliases: |
USP21 |
Location: |
Chr.21:15729947-15880072 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP25 | NM_001283041.1 | NP_001269970.1 | ||
| NM_001283042.1 | NP_001269971.1 | |||
| NM_013396.4 | NP_037528.3 | |||
| XM_011529529.1 | XP_011527831.1 | |||
| XM_011529531.2 | XP_011527833.1 | |||
| XM_011529535.2 | XP_011527837.1 | |||
| XM_017028322.1 | XP_016883811.1 | |||
| XM_017028324.1 | XP_016883813.1 | |||
| XM_017028325.1 | XP_016883814.1 | |||
| XM_017028328.1 | XP_016883817.1 | |||
| AF134213.1 | AAF24998.1 | |||
| AF170562.1 | AAF32263.1 | |||
| BC015930.1 | AAH15930.1 | |||
| BC075792.1 | AAH75792.1 | |||
| FJ763652.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4390n100 | Chr.21:15582251 - 15773439 on Build GRCh38 | Gain |
|
| esv2762093 | Chr.21:15576344 - 15773451 on Build GRCh38 | Gain |
|
| nsv459099 | Chr.21:15584547 - 15772692 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map