Assay Details
Target Gene Details
Entrez Gene ID: | 8624 |
Gene Name: | proteasome assembly chaperone 1 |
Gene Aliases: |
C21LRP, DSCR2, LRPC21, PAC-1, PAC1 |
Location: |
Chr.21:39175446-39183514 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMG1 | NM_001261824.1 | NP_001248753.1 | ||
| NM_001320795.1 | NP_001307724.1 | |||
| NM_003720.3 | NP_003711.1 | |||
| NM_203433.2 | NP_982257.1 | |||
| NR_049728.1 | ||||
| XM_017028488.1 | XP_016883977.1 | |||
| AA731200.1 | ||||
| AB451468.1 | ||||
| AF417108.1 | ||||
| AJ006291.1 | CAA06957.1 | |||
| AK001455.1 | ||||
| AK308614.1 | ||||
| AK315406.1 | ||||
| AW594615.1 | ||||
| AY463963.1 | AAR25628.1 | |||
| BC003619.2 | AAH03619.1 | |||
| BC010424.1 | AAH10424.1 | |||
| BC011755.2 | AAH11755.1 | |||
| BC012809.2 | AAH12809.1 | |||
| BM013494.1 | ||||
| BM462074.1 | ||||
| BQ008489.1 | ||||
| CR541821.1 | CAG46620.1 | |||
| CR541852.1 | CAG46650.1 |
More Information
Set Membership: |
|

Genomic Map