Assay Details
Target Gene Details
Entrez Gene ID: | 29104 |
Gene Name: | N-6 adenine-specific DNA methyltransferase 1 (putative) |
Gene Aliases: |
C21orf127, HEMK2, MTQ2, N6AMT, PRED28, m.HsaHemK2P |
Location: |
Chr.21:28571095-28885373 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N6AMT1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79357839] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map