Assay Details
Target Gene Details
Entrez Gene ID: | 4599 |
Gene Name: | MX dynamin like GTPase 1 |
Gene Aliases: |
IFI-78K, IFI78, MX, MxA |
Location: |
Chr.21:41420329-41459214 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834102 | Chr.21:41286267 - 41494006 on Build GRCh38 | Loss |
|
| esv3893413 | Chr.21:41410704 - 41454624 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map