Assay Details
Target Gene Details
Entrez Gene ID: | 1826 |
Gene Name: | DS cell adhesion molecule |
Gene Aliases: |
CHD2, CHD2-42, CHD2-52 |
Location: |
Chr.21:40010999-40847113 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSCAM | NM_001271534.1 | NP_001258463.1 | ||
| NM_001389.3 | NP_001380.2 | |||
| NR_073202.1 | ||||
| AB209136.1 | BAD92373.1 | |||
| AF023449.1 | AAC17966.1 | |||
| AF023450.1 | AAC17967.1 | |||
| AF217525.1 | AAF27525.1 |
Target Gene Details
Entrez Gene ID: | 100506492 |
Gene Name: | DSCAM antisense RNA 1 |
Gene Aliases: |
M41 |
Location: |
Chr.21:40383083-40385358 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSCAM-AS1 | NR_038896.1 | |||
| NR_038898.1 | 1 | 432 | ||
| NR_038899.1 | ||||
| NR_038900.1 | ||||
| AF401029.1 | 1 | 432 | ||
| AF401030.1 | ||||
| AF401031.1 | ||||
| AF401032.1 | ||||
| AF401033.1 | ||||
| AF401034.1 | ||||
| AF401035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528658 | Chr.21:40371712 - 40388490 on Build GRCh38 | Loss |
|
| nsv520586 | Chr.21:40373074 - 40383482 on Build GRCh38 | Loss |
|
| nsv834100 | Chr.21:40353262 - 40571232 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map