Assay Details
Target Gene Details
Entrez Gene ID: | 56246 |
Gene Name: | melanocortin 2 receptor accessory protein |
Gene Aliases: |
B27, C21orf61, FALP, FGD2, GCCD2 |
Location: |
Chr.21:32291813-32314784 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRAP | NM_001285394.1 | NP_001272323.1 | ||
| NM_178817.3 | NP_848932.1 | |||
| NM_206898.1 | NP_996781.1 | |||
| XM_006724028.3 | XP_006724091.1 | |||
| XM_017028407.1 | XP_016883896.1 | |||
| AF454915.1 | AAL51048.1 | |||
| AF454916.1 | AAL51049.1 | |||
| AF483549.1 | AAL86908.1 | |||
| AY079152.1 | AAL80042.1 | |||
| BC062721.1 | AAH62721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510795 | Chr.21:32272735 - 32382310 on Build GRCh38 | Deletion |
|
| nsv834084 | Chr.21:32179709 - 32380619 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map