Assay Details
Target Gene Details
Entrez Gene ID: | 25825 |
Gene Name: | beta-site APP-cleaving enzyme 2 |
Gene Aliases: |
AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAP |
Location: |
Chr.21:41167801-41282534 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BACE2 | NM_012105.4 | NP_036237.2 | ||
| NM_138991.2 | NP_620476.1 | |||
| NM_138992.2 | NP_620477.1 | |||
| AF050171.1 | AAD45963.1 | |||
| AF117892.1 | AAD45240.1 | |||
| AF178532.1 | AAF29494.1 | |||
| AF188276.1 | AAF35835.1 | |||
| AF188277.1 | AAF35836.1 | |||
| AF200192.1 | AAF13714.1 | |||
| AF200342.1 | AAF17078.1 | |||
| AF204944.1 | AAF26368.1 | |||
| AF212252.1 | AAG41783.1 | |||
| AK075539.1 | BAC11682.1 | |||
| AK292056.1 | ||||
| AY358927.1 | AAQ89286.1 | |||
| BC014453.1 | AAH14453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1144002 | Chr.21:41167073 - 41169173 on Build GRCh38 | Deletion |
|
| nsv1072179 | Chr.21:41167772 - 41169173 on Build GRCh38 | Deletion |
|
| nsv587492 | Chr.21:41137896 - 41175967 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map