Assay Details
Target Gene Details
Entrez Gene ID: | 4599 |
Gene Name: | MX dynamin like GTPase 1 |
Gene Aliases: |
IFI-78K, IFI78, MX, MxA |
Location: |
Chr.21:41420329-41459214 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MX1 | NM_001144925.2 | NP_001138397.1 | ||
| XM_011529568.2 | XP_011527870.1 | |||
| XM_017028349.1 | XP_016883838.1 | |||
| AK096355.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834102 | Chr.21:41286267 - 41494006 on Build GRCh38 | Loss |
|
| esv1009381 | Chr.21:41420102 - 41431317 on Build GRCh38 | Deletion |
|
| esv3893413 | Chr.21:41410704 - 41454624 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map