Assay Details
Target Gene Details
Entrez Gene ID: | 861 |
Gene Name: | runt related transcription factor 1 |
Gene Aliases: |
AML1, AML1-EVI-1, AMLCR1, CBF2alpha, CBFA2, EVI-1, PEBP2aB, PEBP2alpha |
Location: |
Chr.21:34787801-35049334 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUNX1 | NM_001001890.2 | NP_001001890.1 | ||
| NM_001754.4 | NP_001745.2 | |||
| XM_005261068.3 | XP_005261125.1 | |||
| XM_005261069.4 | XP_005261126.1 | |||
| XM_011529766.2 | XP_011528068.1 | |||
| XM_011529767.2 | XP_011528069.1 | |||
| XM_011529768.2 | XP_011528070.1 | |||
| XM_017028487.1 | XP_016883976.1 | |||
| AK226159.1 | ||||
| AL581043.3 | ||||
| BC136380.1 | ||||
| BC136381.1 | ||||
| BC144053.1 | ||||
| D43968.1 | BAA07903.1 | |||
| D43969.1 | BAA07904.1 | |||
| D89788.1 | BAA14020.1 | |||
| D89789.1 | BAA14021.1 | |||
| D89790.1 | BAA14022.1 | |||
| L34598.1 | AAA51720.1 | |||
| U19601.1 | AAB51691.1 | |||
| X79549.1 | CAA56092.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3568160 | Chr.21:34797680 - 34798922 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map