Assay Details
Target Gene Details
Entrez Gene ID: | 3455 |
Gene Name: | interferon alpha and beta receptor subunit 2 |
Gene Aliases: |
IFN-R, IFN-alpha-REC, IFNABR, IFNARB, IMD45 |
Location: |
Chr.21:33229895-33264513 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFNAR2 | NM_000874.4 | NP_000865.2 | ||
| NM_001289125.1 | NP_001276054.1 | |||
| NM_001289126.1 | NP_001276055.1 | |||
| NM_001289128.1 | NP_001276057.1 | |||
| NM_207584.2 | NP_997467.1 | |||
| NM_207585.2 | NP_997468.1 | |||
| BC002793.2 | AAH02793.1 | |||
| BG715143.1 | ||||
| CD110317.1 | ||||
| HG380509.1 | ||||
| HY104613.1 | ||||
| L41942.1 | AAB46413.1 | |||
| L41944.1 | AAB46415.1 | |||
| U29584.1 | AAC50202.1 | |||
| X77722.1 | CAA54785.1 | |||
| X89814.1 | CAA61940.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs189110596] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map