Assay Details
Target Gene Details
Entrez Gene ID: | 157 |
Gene Name: | G protein-coupled receptor kinase 3 |
Gene Aliases: |
ADRBK2, BARK2 |
Location: |
Chr.22:25564894-25729294 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK3 | NM_005160.3 | NP_005151.2 | ||
| AK058103.1 | ||||
| AK292234.1 | ||||
| AK310398.1 | ||||
| BC036797.1 | AAH36797.1 | |||
| BX387339.2 | ||||
| X69117.1 | CAA48870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066595 | Chr.22:25596614 - 26071416 on Build GRCh38 | Gain |
|
| nsv517205 | Chr.22:25244661 - 25629591 on Build GRCh38 | Gain+Loss |
|
| esv2758829 | Chr.22:24970834 - 25650546 on Build GRCh38 | Gain+Loss |
|
| nsv1058343 | Chr.22:25507736 - 25642811 on Build GRCh38 | Loss |
|
| esv2751952 | Chr.22:25262579 - 25754679 on Build GRCh38 | Gain |
|
| esv3647478 | Chr.22:25598397 - 25702971 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map