Assay Details
Target Gene Details
Entrez Gene ID: | 27442 |
Gene Name: | cat eye syndrome chromosome region, candidate 3 (non-protein coding) |
Gene Aliases: |
- |
Location: |
Chr.22:17256859-17266733 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CECR3 | NR_038398.2 | 3 | 857 | |
| AF277398.1 | 3 | 857 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647209 | Chr.22:17151829 - 17264992 on Build GRCh38 | Gain |
|
| nsv1067241 | Chr.22:16757380 - 17351252 on Build GRCh38 | Loss |
|
| esv3647213 | Chr.22:17256922 - 17261112 on Build GRCh38 | Gain |
|
| esv3893427 | Chr.22:17214813 - 17405801 on Build GRCh38 | Gain |
|
| nsv509805 | Chr.22:17081417 - 17267685 on Build GRCh38 | Insertion |
|

Genomic Map