Assay Details
Target Gene Details
Entrez Gene ID: | 27443 |
Gene Name: | CECR2, histone acetyl-lysine reader |
Gene Aliases: |
- |
Location: |
Chr.22:17359949-17558155 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CECR2 | NM_001290046.1 | NP_001276975.1 | ||
| AK308330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv513582 | Chr.22:17368440 - 17370586 on Build GRCh38 | Insertion |
|
| nsv459335 | Chr.22:17333921 - 17436386 on Build GRCh38 | Gain |
|
| nsv834118 | Chr.22:17358660 - 17523783 on Build GRCh38 | Loss |
|
| esv3893427 | Chr.22:17214813 - 17405801 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73876414] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map