Assay Details
Target Gene Details
Entrez Gene ID: | 27350 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 3C |
Gene Aliases: |
A3C, APOBEC1L, ARDC2, ARDC4, ARP5, PBI, bK150C2.3 |
Location: |
Chr.22:39014260-39018838 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOBEC3C | NM_014508.2 | NP_055323.2 | ||
| AB208979.1 | BAD92216.1 | |||
| AF165520.1 | AAF86650.1 | |||
| AK313272.1 | ||||
| BC011739.2 | AAH11739.1 | |||
| CR456394.1 | CAG30280.1 | |||
| KC329528.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv962793 | Chr.22:39014160 - 39017969 on Build GRCh38 | Duplication |
|
| esv2763703 | Chr.22:38955461 - 39034450 on Build GRCh38 | Loss |
|
| esv2724241 | Chr.22:39014940 - 39028280 on Build GRCh38 | Deletion |
|
| esv3647751 | Chr.22:38997523 - 39050430 on Build GRCh38 | Gain |
|
| esv3647753 | Chr.22:39004677 - 39029340 on Build GRCh38 | Gain |
|
| nsv834196 | Chr.22:38996564 - 39214999 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs146055408] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map