Assay Details
Target Gene Details
Entrez Gene ID: | 266629 |
Gene Name: | SEC14 like lipid binding 3 |
Gene Aliases: |
TAP2 |
Location: |
Chr.22:30447959-30472047 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC14L3 | XM_011530128.2 | 12 | 2309 | XP_011528430.1 |
| AK131358.1 | 13 | 2363 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588886 | Chr.22:30440326 - 30472431 on Build GRCh38 | Loss |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map