Assay Details
Target Gene Details
Entrez Gene ID: | 5155 |
Gene Name: | platelet derived growth factor subunit B |
Gene Aliases: |
IBGC5, PDGF-2, PDGF2, SIS, SSV, c-sis |
Location: |
Chr.22:39223359-39245055 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDGFB | NM_002608.3 | NP_002599.1 | ||
| NM_033016.3 | NP_148937.1 | |||
| BC029822.1 | AAH29822.1 | |||
| BC077725.1 | AAH77725.1 | |||
| CR456538.1 | CAG30424.1 | |||
| CR541807.1 | CAG46606.1 | |||
| M12783.1 | AAA60553.1 | |||
| X02744.1 | CAA26524.1 | |||
| X02811.1 | CAA26579.1 | |||
| X03702.1 | CAA27333.1 | |||
| X83705.1 | CAA58679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647761 | Chr.22:39204243 - 39286742 on Build GRCh38 | Gain |
|
| nsv1160762 | Chr.22:39214849 - 39298220 on Build GRCh38 | Deletion |
|
| nsv834198 | Chr.22:39222748 - 39409818 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map