Assay Details
Target Gene Details
Entrez Gene ID: | 5435 |
Gene Name: | RNA polymerase II subunit F |
Gene Aliases: |
HRBP14.4, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPC15 |
Location: |
Chr.22:37953663-38041915 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2F | NM_001301130.1 | NP_001288059.1 | ||
| NM_001301131.1 | NP_001288060.1 | |||
| XM_017028832.1 | XP_016884321.1 | |||
| AK094034.1 | ||||
| BP195499.1 | ||||
| DN989996.1 |
Target Gene Details
Entrez Gene ID: | 6663 |
Gene Name: | SRY-box 10 |
Gene Aliases: |
DOM, PCWH, WS2E, WS4, WS4C |
Location: |
Chr.22:37972312-37984532 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX10 | NM_006941.3 | NP_008872.1 | ||
| AJ001183.1 | CAA04576.1 | |||
| AK300945.1 | ||||
| BC002824.2 | AAH02824.1 | |||
| BC007595.1 | AAH07595.1 | |||
| BT020029.1 | AAV38832.1 | |||
| CR456584.1 | CAG30470.1 | |||
| CR536571.1 | CAG38808.1 |
More Information
Set Membership: |
|

Genomic Map