Assay Details
Target Gene Details
Entrez Gene ID: | 100506195 |
Gene Name: | LARGE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.22:33724986-33750817 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARGE-AS1 | NR_038949.1 | |||
| NR_038950.1 |
Target Gene Details
Entrez Gene ID: | 9215 |
Gene Name: | LARGE xylosyl- and glucuronyltransferase 1 |
Gene Aliases: |
LARGE, MDC1D, MDDGA6, MDDGB6 |
Location: |
Chr.22:33162237-33922841 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARGE1 | NM_004737.4 | NP_004728.1 | ||
| NM_133642.3 | NP_598397.1 | |||
| XM_005261831.3 | XP_005261888.1 | |||
| XM_005261832.3 | XP_005261889.1 | |||
| XM_011530510.2 | XP_011528812.1 | |||
| XM_011530514.2 | XP_011528816.1 | |||
| AB011181.3 | BAA25535.3 | |||
| AJ007583.1 | CAA07571.1 | |||
| AK294766.1 | ||||
| AK295636.1 | ||||
| BC117425.1 | ||||
| BC126404.1 | ||||
| CR456510.1 | CAG30396.1 | |||
| KJ534877.1 | ||||
| KJ535033.1 | ||||
| KJ535073.1 | ||||
| KJ535074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055791 | Chr.22:33426240 - 33763549 on Build GRCh38 | Loss |
|
| nsv524432 | Chr.22:33723880 - 33750381 on Build GRCh38 | Loss |
|
| nsv588936 | Chr.22:33710759 - 33830604 on Build GRCh38 | Loss |
|
| esv2763261 | Chr.22:33435362 - 33892401 on Build GRCh38 | Loss |
|
| esv3568379 | Chr.22:33458829 - 33815605 on Build GRCh38 | Loss |
|
| nsv1055203 | Chr.22:33702431 - 33800698 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map