Assay Details
Target Gene Details
Entrez Gene ID: | 10478 |
Gene Name: | solute carrier family 25 member 17 |
Gene Aliases: |
PMP34 |
Location: |
Chr.22:40769630-40819399 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A17 | NM_001282726.1 | NP_001269655.1 | ||
| NM_001282727.1 | NP_001269656.1 | |||
| NM_006358.3 | NP_006349.1 | |||
| NR_104235.1 | ||||
| NR_104236.1 | ||||
| NR_104237.1 | ||||
| NR_104238.1 | ||||
| AK094062.1 | ||||
| AK292924.1 | ||||
| AK298215.1 | ||||
| AK300553.1 | ||||
| AU123445.1 | ||||
| BC005957.1 | AAH05957.1 | |||
| BC012998.2 | AAH12998.1 | |||
| BQ440957.1 | ||||
| BU595416.1 | ||||
| BX352732.2 | ||||
| BX647991.1 | ||||
| CR456577.1 | CAG30463.1 | |||
| Y12860.1 | CAA73367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834206 | Chr.22:40696722 - 40838881 on Build GRCh38 | Gain |
|
| nsv1062438 | Chr.22:40646389 - 40806134 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map