Assay Details
Target Gene Details
Entrez Gene ID: | 140564 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 3D |
Gene Aliases: |
A3D, APOBEC3DE, APOBEC3E, ARP6 |
Location: |
Chr.22:39021113-39034328 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOBEC3D | NM_152426.3 | NP_689639.2 | ||
| XM_005261337.3 | XP_005261394.1 | |||
| XM_017028596.1 | XP_016884085.1 | |||
| BE888971.1 | ||||
| CR456396.1 | CAG30282.1 | |||
| EU527884.1 |
Target Gene Details
Entrez Gene ID: | 200316 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 3F |
Gene Aliases: |
A3F, ARP8, BK150C2.4.MRNA, KA6 |
Location: |
Chr.22:39020454-39055972 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOBEC3F | XM_017028643.1 | XP_016884132.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763703 | Chr.22:38955461 - 39034450 on Build GRCh38 | Loss |
|
| esv2724241 | Chr.22:39014940 - 39028280 on Build GRCh38 | Deletion |
|
| esv3647751 | Chr.22:38997523 - 39050430 on Build GRCh38 | Gain |
|
| esv3647753 | Chr.22:39004677 - 39029340 on Build GRCh38 | Gain |
|
| nsv834196 | Chr.22:38996564 - 39214999 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map