Assay Details
Target Gene Details
Entrez Gene ID: | 101927257 |
Gene Name: | uncharacterized LOC101927257 |
Gene Aliases: |
- |
Location: |
Chr.22:40521800-40526707 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927257 | NR_109965.1 | |||
| AK097769.1 |
Target Gene Details
Entrez Gene ID: | 57591 |
Gene Name: | megakaryoblastic leukemia (translocation) 1 |
Gene Aliases: |
BSAC, MAL, MRTF-A |
Location: |
Chr.22:40410281-40636719 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MKL1 | NM_001282661.1 | NP_001269590.1 | ||
| NM_001282662.1 | NP_001269591.1 | |||
| NM_001318139.1 | NP_001305068.1 | |||
| NM_020831.4 | NP_065882.1 | |||
| XM_005261692.1 | XP_005261749.1 | |||
| XM_005261694.1 | XP_005261751.1 | |||
| XM_011530283.1 | XP_011528585.1 | |||
| XM_011530285.1 | XP_011528587.1 | |||
| XM_017028888.1 | XP_016884377.1 | |||
| AB037859.2 | ||||
| AF368061.1 | AAK54721.1 | |||
| AF448806.1 | ||||
| AJ297258.1 | CAC38827.1 | |||
| BC114364.2 | AAI14365.1 | |||
| BC115039.1 | ||||
| HG764171.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv771n67 | Chr.22:40486132 - 40568329 on Build GRCh38 | Gain |
|
| nsv589047 | Chr.22:40525676 - 40679539 on Build GRCh38 | Loss |
|
| nsv1160763 | Chr.22:40515733 - 40728482 on Build GRCh38 | Duplication |
|
| esv3568411 | Chr.22:40520792 - 40530718 on Build GRCh38 | Loss |
|
| nsv589046 | Chr.22:40490740 - 40600363 on Build GRCh38 | Loss |
|
| nsv829234 | Chr.22:40480449 - 40567496 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map