Assay Details
Target Gene Details
Entrez Gene ID: | 101929664 |
Gene Name: | HORMAD2 antisense RNA 1 |
Gene Aliases: |
MTMR3-AS1 |
Location: |
Chr.22:30008742-30080480 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HORMAD2-AS1 | NR_110541.1 | |||
| BG618381.1 |
Target Gene Details
Entrez Gene ID: | 8897 |
Gene Name: | myotubularin related protein 3 |
Gene Aliases: |
FYVE-DSP1, ZFYVE10 |
Location: |
Chr.22:29883165-30030868 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
More Information
Set Membership: |
|

Genomic Map