Assay Details
Target Gene Details
Entrez Gene ID: | 100423034 |
Gene Name: | microRNA 3199-1 |
Gene Aliases: |
mir-3199-1 |
Location: |
Chr.22:27920525-27920612 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR3199-1 | NR_036169.1 |
Target Gene Details
Entrez Gene ID: | 100422998 |
Gene Name: | microRNA 3199-2 |
Gene Aliases: |
mir-3199-2 |
Location: |
Chr.22:27920526-27920611 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR3199-2 | NR_036170.1 |
Target Gene Details
Entrez Gene ID: | 284900 |
Gene Name: | TTC28 antisense RNA 1 |
Gene Aliases: |
TTC28-AS, TTC28AS, dJ353E16.2 |
Location: |
Chr.22:27919376-28002679 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC28-AS1 | NR_026962.1 | |||
| NR_026963.1 | ||||
| AL832597.1 | ||||
| DA934139.1 | ||||
| DB254752.1 |

Genomic Map