Assay Details
Target Gene Details
Entrez Gene ID: | 55687 |
Gene Name: | tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase |
Gene Aliases: |
LCAL3, MTO2, MTU1, TRMT, TRMT1, TRNT1 |
Location: |
Chr.22:46335401-46357340 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829293 | Chr.22:46241054 - 46741639 on Build GRCh38 | Gain |
|
| nsv829281 | Chr.22:45861485 - 46735858 on Build GRCh38 | Gain |
|
| nsv834225 | Chr.22:46323584 - 46510799 on Build GRCh38 | Loss |
|
| nsv471214 | Chr.22:46278315 - 46367774 on Build GRCh38 | Loss |
|
| nsv1066139 | Chr.22:46310352 - 46352592 on Build GRCh38 | Gain |
|
| esv3647951 | Chr.22:46324981 - 46344914 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114761740] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map