Assay Details
Target Gene Details
Entrez Gene ID: | 412 |
Gene Name: | steroid sulfatase (microsomal), isozyme S |
Gene Aliases: |
ARSC, ARSC1, ASC, ES, SSDD, XLI |
Location: |
Chr.X:7147252-7354643 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STS | NM_001320750.1 | NP_001307679.1 | ||
| NM_001320751.1 | NP_001307680.1 | |||
| NM_001320752.1 | NP_001307681.1 | |||
| AM072428.1 | ||||
| AM072429.1 | ||||
| AM282551.1 | ||||
| DA327298.1 | ||||
| DA406759.1 | ||||
| FM209047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576734 | Chr.X:6537111 - 7202526 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map