Assay Details
Target Gene Details
Entrez Gene ID: | 11238 |
Gene Name: | carbonic anhydrase 5B |
Gene Aliases: |
CA-VB |
Location: |
Chr.X:15738281-15787625 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA5B | NM_007220.3 | NP_009151.1 | ||
| XM_005274442.4 | XP_005274499.1 | |||
| XM_017029249.1 | XP_016884738.1 | |||
| AB021660.1 | BAA76671.1 | |||
| AK291050.1 | ||||
| BP371510.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739978 | Chr.X:15696314 - 15775807 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map