Assay Details
Target Gene Details
Entrez Gene ID: | 1184 |
Gene Name: | chloride voltage-gated channel 5 |
Gene Aliases: |
CLC5, CLCK2, ClC-5, DENTS, NPHL1, NPHL2, XLRH, XRN, hCIC-K2 |
Location: |
Chr.X:49922615-50099235 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLCN5 | NM_001127898.3 | NP_001121370.1 | ||
| NM_001127899.3 | NP_001121371.1 | |||
| NM_001272102.1 | NP_001259031.1 | |||
| XM_017029257.1 | XP_016884746.1 | |||
| XM_017029258.1 | XP_016884747.1 | |||
| AK056560.1 | ||||
| AK092049.1 | ||||
| BF513571.1 |
Target Gene Details
Entrez Gene ID: | 724030 |
Gene Name: | microRNA 660 |
Gene Aliases: |
MIRN660, hsa-mir-660, mir-660 |
Location: |
Chr.X:50013241-50013337 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR660 | NR_030397.1 |
More Information
Set Membership: |
|

Genomic Map