Assay Details
Target Gene Details
Entrez Gene ID: | 59272 |
Gene Name: | angiotensin I converting enzyme 2 |
Gene Aliases: |
ACEH |
Location: |
Chr.X:15494525-15602069 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACE2 | XM_011545549.1 | XP_011543851.1 | ||
| XM_011545551.2 | XP_011543853.1 |
Target Gene Details
Entrez Gene ID: | 660 |
Gene Name: | BMX non-receptor tyrosine kinase |
Gene Aliases: |
ETK, PSCTK2, PSCTK3 |
Location: |
Chr.X:15500777-15556529 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BMX | NM_001320866.1 | NP_001307795.1 | ||
| NM_001721.6 | NP_001712.1 | |||
| NM_203281.2 | NP_975010.1 | |||
| XM_017029752.1 | XP_016885241.1 | |||
| AF045459.1 | AAC08966.1 | |||
| AK291824.1 | ||||
| AK313568.1 | ||||
| BC016652.2 | AAH16652.1 | |||
| BQ226991.1 | ||||
| X83107.1 | CAA58169.1 |
More Information
Set Membership: |
|

Genomic Map