Assay Details
Target Gene Details
Entrez Gene ID: | 30011 |
Gene Name: | SH3 domain containing kinase binding protein 1 |
Gene Aliases: |
CD2BP3, CIN85, GIG10, HSB-1, HSB1, MIG18 |
Location: |
Chr.X:19533965-19887626 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6824 | Chr.X:19536753 - 19582095 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map