Assay Details
Target Gene Details
Entrez Gene ID: | 79979 |
Gene Name: | tRNA methyltransferase 2 homolog B |
Gene Aliases: |
CXorf34, dJ341D10.3 |
Location: |
Chr.X:100973358-101052118 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT2B |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517937 | Chr.X:100982949 - 101015369 on Build GRCh38 | Gain |
|
| nsv515758 | Chr.X:100987316 - 101038566 on Build GRCh38 | Gain |
|
| nsv7013 | Chr.X:100994802 - 101039808 on Build GRCh38 | Deletion |
|
| nsv7012 | Chr.X:100965059 - 100999697 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs143303815] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map