Assay Details
Target Gene Details
Entrez Gene ID: | 79868 |
Gene Name: | ALG13, UDP-N-acetylglucosaminyltransferase subunit |
Gene Aliases: |
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W |
Location: |
Chr.X:111680741-111764164 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
More Information
Set Membership: |
|

Genomic Map