Assay Details
Target Gene Details
Entrez Gene ID: | 29071 |
Gene Name: | C1GALT1 specific chaperone 1 |
Gene Aliases: |
C1GALT2, C38H2-L1, COSMC, HSPC067, MST143, TNPS |
Location: |
Chr.X:120625674-120630150 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1GALT1C1 | NM_001011551.2 | NP_001011551.1 | ||
| NM_152692.4 | NP_689905.1 | |||
| AB084170.1 | BAC41493.1 | |||
| AF161552.1 | AAF29039.1 | |||
| AF177284.1 | AAQ13670.1 | |||
| AJ238398.1 | CAC80277.1 | |||
| AK290111.1 | ||||
| AY159319.1 | AAN78129.1 | |||
| AY358642.1 | AAQ89005.1 | |||
| BC011930.2 | AAH11930.1 | |||
| BC050441.1 | AAH50441.1 | |||
| BG779539.1 |
More Information
Set Membership: |
|

Genomic Map