Assay Details
Target Gene Details
Entrez Gene ID: | 6792 |
Gene Name: | cyclin dependent kinase like 5 |
Gene Aliases: |
CFAP247, EIEE2, ISSX, STK9 |
Location: |
Chr.X:18425605-18653629 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKL5 | NM_003159.2 | NP_003150.1 | ||
| AK307444.1 | ||||
| AY217744.1 | AAO64440.1 | |||
| Y15057.1 | CAA75342.1 |
More Information
Set Membership: |
|

Genomic Map