Assay Details
Target Gene Details
Entrez Gene ID: | 1968 |
Gene Name: | eukaryotic translation initiation factor 2 subunit gamma |
Gene Aliases: |
EIF2, EIF2G, EIF2gamma, eIF-2gA |
Location: |
Chr.X:24054948-24078810 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2S3 | NM_001415.3 | NP_001406.1 | ||
| AB451230.1 | ||||
| AB451353.1 | ||||
| AK222577.1 | BAD96297.1 | |||
| AK290397.1 | ||||
| AK312247.1 | ||||
| BC019906.1 | AAH19906.1 | |||
| L19161.1 | AAA19696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524256 | Chr.X:24021660 - 24160925 on Build GRCh38 | Gain |
|
| esv2740010 | Chr.X:23974798 - 24136109 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map