Assay Details
Target Gene Details
Entrez Gene ID: | 389840 |
Gene Name: | mitogen-activated protein kinase kinase kinase 15 |
Gene Aliases: |
ASK3, bA723P2.3 |
Location: |
Chr.X:19360058-19515459 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K15 | NM_001001671.3 | NP_001001671.3 | ||
| XM_011545507.2 | XP_011543809.2 | |||
| XM_011545508.2 | XP_011543810.2 | |||
| XM_011545510.2 | XP_011543812.1 | |||
| XM_011545511.2 | XP_011543813.1 | |||
| AK122632.1 | ||||
| AK131412.1 | BAD18559.1 | |||
| AK131477.1 | BAD18622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573627 | Chr.X:19351701 - 19362562 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map