Assay Details
Target Gene Details
Entrez Gene ID: | 11141 |
Gene Name: | interleukin 1 receptor accessory protein like 1 |
Gene Aliases: |
IL1R8, IL1RAPL, MRX10, MRX21, MRX34, OPHN4, TIGIRR-2 |
Location: |
Chr.X:28587564-29956350 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1RAPL1 | NM_014271.3 | NP_055086.1 | ||
| AF181284.1 | AAF59411.1 | |||
| AF284435.1 | AAG21369.1 | |||
| AJ243874.1 | CAB56046.1 | |||
| DN994486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521056 | Chr.X:28590019 - 28664578 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140875510] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map