Assay Details
Target Gene Details
Entrez Gene ID: | 4935 |
Gene Name: | G protein-coupled receptor 143 |
Gene Aliases: |
NYS6, OA1 |
Location: |
Chr.X:9725413-9765965 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR143 | NM_000273.2 | NP_000264.2 | ||
| XM_005274541.3 | XP_005274598.1 | |||
| BC068977.1 | AAH68977.1 | |||
| Z48804.1 | CAA88742.1 |
More Information
Set Membership: |
|

Genomic Map