Assay Details
Target Gene Details
Entrez Gene ID: | 5354 |
Gene Name: | proteolipid protein 1 |
Gene Aliases: |
GPM6C, HLD1, MMPL, PLP, PLP/DM20, PMD, SPG2 |
Location: |
Chr.X:103776506-103792619 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLP1 | NM_000533.4 | NP_000524.3 | ||
| NM_001128834.2 | NP_001122306.1 | |||
| NM_001305004.1 | NP_001291933.1 | |||
| NM_199478.2 | NP_955772.1 | |||
| AK128782.1 | ||||
| AK292728.1 | ||||
| AK295374.1 | ||||
| AK295388.1 | ||||
| AK309239.1 | ||||
| AK312340.1 | ||||
| BC002665.1 | AAH02665.1 | |||
| BC095452.1 | AAH95452.1 | |||
| BQ717999.1 | ||||
| BX445448.2 | ||||
| CA389667.1 | ||||
| DA299940.1 | ||||
| DC342197.1 | ||||
| DC342996.1 | ||||
| M27110.1 | AAA60117.1 | |||
| M54927.1 | AAA59565.1 |
More Information
Set Membership: |
|

Genomic Map