Assay Details
Target Gene Details
Entrez Gene ID: | 10495 |
Gene Name: | ecto-NOX disulfide-thiol exchanger 2 |
Gene Aliases: |
APK1, COVA1, tNOX |
Location: |
Chr.X:130622330-130903317 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENOX2 | NM_001281736.1 | NP_001268665.1 | ||
| NM_006375.3 | NP_006366.2 | |||
| NM_182314.2 | NP_872114.1 | |||
| XM_005262354.3 | XP_005262411.1 | |||
| XM_011531244.2 | XP_011529546.1 | |||
| XM_011531245.2 | XP_011529547.1 | |||
| XM_011531247.2 | XP_011529549.1 | |||
| XM_011531251.2 | XP_011529553.1 | |||
| XM_017029226.1 | XP_016884715.1 | |||
| XM_017029227.1 | XP_016884716.1 | |||
| XM_017029229.1 | XP_016884718.1 | |||
| AK000353.1 | ||||
| AK289812.1 | ||||
| AK289837.1 | ||||
| AL133207.2 | CAB61581.2 | |||
| BC108713.1 | AAI08714.1 | |||
| BU160341.1 |
Target Gene Details
Entrez Gene ID: | 105373338 |
Gene Name: | uncharacterized LOC105373338 |
Gene Aliases: |
- |
Location: |
Chr.X:130802636-130806398 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373338 | NR_135655.1 | |||
| DB449817.1 | ||||
| HY002605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527144 | Chr.X:130744151 - 131039499 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map