Assay Details
Target Gene Details
Entrez Gene ID: | 5303 |
Gene Name: | peptidylprolyl cis/trans isomerase, NIMA-interacting 4 |
Gene Aliases: |
EPVH, PAR14, PAR17 |
Location: |
Chr.X:72181676-72263964 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIN4 | NM_001170747.1 | NP_001164218.1 | ||
| NM_006223.3 | NP_006214.2 | |||
| NR_033187.1 | ||||
| AB009690.1 | BAA82320.1 | |||
| AF143096.1 | AAD27893.1 | |||
| AK127605.1 | ||||
| AU098526.1 | ||||
| AV709926.2 | ||||
| BC005234.1 | AAH05234.2 | |||
| BC070288.1 | AAH70288.1 | |||
| BC093700.1 | AAH93700.1 | |||
| BC104653.1 | AAI04654.1 | |||
| BC111394.1 | ||||
| BC112281.1 | AAI12282.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2673799 | Chr.X:72189046 - 72196415 on Build GRCh38 | Deletion |
|
| esv21569 | Chr.X:72189299 - 72196211 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73550759] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map