Assay Details
Target Gene Details
Entrez Gene ID: | 4935 |
Gene Name: | G protein-coupled receptor 143 |
Gene Aliases: |
NYS6, OA1 |
Location: |
Chr.X:9725413-9765965 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR143 | NM_000273.2 | NP_000264.2 | ||
| XM_005274541.3 | XP_005274598.1 | |||
| BC068977.1 | AAH68977.1 | |||
| Z48804.1 | CAA88742.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs145554694] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map