Assay Details
Target Gene Details
Entrez Gene ID: | 23597 |
Gene Name: | acyl-CoA thioesterase 9 |
Gene Aliases: |
ACATE2, CGI-16, MT-ACT48, MTACT48 |
Location: |
Chr.X:23701055-23743290 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOT9 | XM_005274472.2 | 14 | 2119 | XP_005274529.1 |
| AF132950.1 | AAD27725.1 | |||
| BX648512.1 | 16 | 2231 |
More Information
Set Membership: |
|

Genomic Map