Assay Details
Target Gene Details
Entrez Gene ID: | 79868 |
Gene Name: | ALG13, UDP-N-acetylglucosaminyltransferase subunit |
Gene Aliases: |
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W |
Location: |
Chr.X:111680741-111764164 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALG13 | XM_006724693.2 | 1 | 82 | XP_006724756.1 |
| XM_006724697.3 | 1 | 86 | XP_006724760.2 | |
| XM_006724698.3 | 1 | 86 | XP_006724761.2 | |
| XM_011531028.2 | 1 | 83 | XP_011529330.1 | |
| XM_011531030.1 | 1 | 81 | XP_011529332.1 | |
| XM_017029846.1 | 1 | 84 | XP_016885335.1 | |
| XM_017029847.1 | 1 | 83 | XP_016885336.1 | |
| XM_017029848.1 | 1 | 81 | XP_016885337.1 | |
| XM_017029849.1 | 1 | 82 | XP_016885338.1 | |
| XM_017029850.1 | 1 | 81 | XP_016885339.1 | |
| XM_017029851.1 | 1 | 84 | XP_016885340.1 |
More Information
Set Membership: |
|

Genomic Map