Assay Details
Target Gene Details
Entrez Gene ID: | 26523 |
Gene Name: | argonaute 1, RISC catalytic component |
Gene Aliases: |
EIF2C, EIF2C1, GERP95, Q99, hAgo1 |
Location: |
Chr.1:35869808-35929610 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGO1 | NM_001317122.1 | NP_001304051.1 | ||
| NM_001317123.1 | NP_001304052.1 | |||
| NM_012199.4 | NP_036331.1 | |||
| XM_011541236.2 | XP_011539538.1 | |||
| XM_017001016.1 | XP_016856505.1 | |||
| XM_017001017.1 | XP_016856506.1 | |||
| XM_017001019.1 | XP_016856508.1 | |||
| AF093097.1 | AAF00068.1 | |||
| AK001622.1 | ||||
| AK314148.1 | ||||
| BC063275.1 | AAH63275.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834680 | Chr.1:35660621 - 35892216 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map