Assay Details
Target Gene Details
Entrez Gene ID: | 23499 |
Gene Name: | microtubule-actin crosslinking factor 1 |
Gene Aliases: |
ABP620, ACF7, MACF, OFC4 |
Location: |
Chr.1:39084167-39487138 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MACF1 | NM_012090.5 | NP_036222.3 | ||
| XM_006710526.3 | XP_006710589.1 | |||
| XM_017000875.1 | XP_016856364.1 | |||
| XM_017000881.1 | XP_016856370.1 | |||
| AB029290.1 | BAA83821.1 | |||
| AK299010.1 | ||||
| BX107650.1 | ||||
| DA512687.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75028003] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map