Assay Details
Target Gene Details
| Entrez Gene ID: | 23028 | 
| Gene Name: | lysine demethylase 1A | 
| Gene Aliases: | AOF2, BHC110, CPRF, KDM1, LSD1 | 
| Location: | Chr.1:23019443-23083691 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| KDM1A | NM_001009999.2 | NP_001009999.1 | ||
| NM_015013.3 | NP_055828.2 | |||
| XM_005245786.2 | XP_005245843.1 | |||
| XM_006710472.3 | XP_006710535.1 | |||
| XM_006710473.2 | XP_006710536.1 | |||
| XM_006710474.3 | XP_006710537.1 | |||
| XM_017000716.1 | XP_016856205.1 | |||
| XM_017000717.1 | XP_016856206.1 | |||
| XM_017000718.1 | XP_016856207.1 | |||
| XM_017000719.1 | XP_016856208.1 | |||
| XM_017000720.1 | XP_016856209.1 | |||
| AB011173.1 | ||||
| AK290328.1 | ||||
| BC040194.2 | AAH40194.3 | |||
| BC048134.1 | AAH48134.2 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  non-DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top