Assay Details
Target Gene Details
Entrez Gene ID: | 59352 |
Gene Name: | leucine rich repeat containing G protein-coupled receptor 6 |
Gene Aliases: |
GPCR, VTS20631 |
Location: |
Chr.1:202193990-202319761 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LGR6 | NM_001017403.1 | NP_001017403.1 | ||
| NM_021636.2 | NP_067649.2 | |||
| AB049405.1 | BAB39854.1 | |||
| AY358119.1 | AAQ88486.1 | |||
| BC047905.1 | AAH47905.2 | |||
| BM699180.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010102 | Chr.1:202164648 - 202260980 on Build GRCh38 | Gain |
|
| nsv1014912 | Chr.1:202206757 - 202242605 on Build GRCh38 | Gain |
|
| nsv522841 | Chr.1:202150606 - 202311878 on Build GRCh38 | Gain |
|
| esv3891059 | Chr.1:202184455 - 202231671 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map