Assay Details
Target Gene Details
Entrez Gene ID: | 79033 |
Gene Name: | ERI1 exoribonuclease family member 3 |
Gene Aliases: |
PINT1, PRNPIP |
Location: |
Chr.1:44221070-44355298 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERI3 | NM_001301698.1 | NP_001288627.1 | ||
| NM_001301699.1 | NP_001288628.1 | |||
| NM_001301700.1 | NP_001288629.1 | |||
| NM_001301701.1 | NP_001288630.1 | |||
| NM_024066.2 | NP_076971.1 | |||
| XM_005271186.2 | XP_005271243.1 | |||
| XM_006710891.3 | XP_006710954.1 | |||
| XM_017002301.1 | XP_016857790.1 | |||
| XM_017002302.1 | XP_016857791.1 | |||
| XM_017002303.1 | XP_016857792.1 | |||
| XM_017002304.1 | XP_016857793.1 | |||
| XM_017002305.1 | XP_016857794.1 | |||
| AF007157.1 | AAC19158.1 | |||
| AK026596.1 | ||||
| AK290567.1 | ||||
| AK293833.1 | ||||
| AK297708.1 | ||||
| BC001072.2 | AAH01072.1 | |||
| BC004456.1 | AAH04456.1 | |||
| BI765887.1 |
Target Gene Details
Entrez Gene ID: | 100874278 |
Gene Name: | ERI3 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.1:44222595-44244283 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERI3-IT1 | NR_110056.1 | |||
| DB453456.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv998002 | Chr.1:44230032 - 44235866 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map