Assay Details
Target Gene Details
Entrez Gene ID: | 55924 |
Gene Name: | family with sequence similarity 212 member B |
Gene Aliases: |
C1orf183 |
Location: |
Chr.1:111722064-111755797 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM212B | NM_198926.2 | NP_945120.1 | ||
| AK293989.1 | ||||
| AL137199.1 | CAB69909.1 |
Target Gene Details
Entrez Gene ID: | 100506343 |
Gene Name: | FAM212B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:111739841-111747798 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM212B-AS1 | NR_038951.1 | |||
| NR_038952.1 | ||||
| AK092511.1 | ||||
| BG329450.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114271578] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map