Assay Details
Target Gene Details
Entrez Gene ID: | 23266 |
Gene Name: | adhesion G protein-coupled receptor L2 |
Gene Aliases: |
CIRL2, CL2, LEC1, LPHH1, LPHN2 |
Location: |
Chr.1:81306155-81992738 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRL2 | XM_017000782.1 | XP_016856271.1 | ||
| AK123422.1 |
Target Gene Details
Entrez Gene ID: | 101927434 |
Gene Name: | uncharacterized LOC101927434 |
Gene Aliases: |
- |
Location: |
Chr.1:81513880-81557702 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927434 | NR_125943.1 | |||
| BX092247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2557027 | Chr.1:81514048 - 81514168 on Build GRCh38 | Deletion |
|
| esv2029393 | Chr.1:81513829 - 81514385 on Build GRCh38 | Deletion |
|
| nsv955454 | Chr.1:81514046 - 81514204 on Build GRCh38 | Deletion |
|
| esv2713919 | Chr.1:81513866 - 81514435 on Build GRCh38 | Deletion |
|
| esv4235 | Chr.1:81513913 - 81514431 on Build GRCh38 | Loss |
|
| esv2464487 | Chr.1:81513454 - 81514874 on Build GRCh38 | Deletion |
|
| esv2763570 | Chr.1:81470555 - 81576665 on Build GRCh38 | Gain |
|
| esv2713930 | Chr.1:81514054 - 81514170 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map